Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Thermo Scientific Chemicals

Potassium nitrate, 99+%, ACS reagent

CAS: 7757-79-1 | KNO3 | 101.10 g/mol
数量:
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 424150025
価格(JPY)
-
数量:
2.5 kg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7757-79-1
IUPAC Namepotassium nitrate
Molecular FormulaKNO3
InChI KeyFGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES[K+].[O-][N+]([O-])=O
さらに表示
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Nitrite (NO2)=<0.001 %
Sulfate (SO4)=<0.003 %
Heavy metals (ICP-OES)=<5 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only