1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

1-Propynylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 16466-97-0 | C3H3BrMg | 143.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number 426071000
Price (USD)
122.65
Special offer
Online exclusive
Ends: 28-Dec-2026
144.00
Save 21.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
122.65
Special offer
Online exclusive
Ends: 28-Dec-2026
144.00
Save 21.35
Each
Chemical Identifiers
CAS16466-97-0
IUPAC Namebromo(prop-1-yn-1-yl)magnesium
Molecular FormulaC3H3BrMg
InChI KeyYVJBCTFVENNIOV-UHFFFAOYSA-M
SMILESCC#C[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
AppearanceMay develop some turbidity or precipitate
Titration Grignard0.40 to 0.60 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only