2-Bromo-2'-chloroacetophenone, 95%
2-Bromo-2'-chloroacetophenone, 95%
2-Bromo-2'-chloroacetophenone, 95%
Thermo Scientific Chemicals

2-Bromo-2'-chloroacetophenone, 95%

CAS: 5000-66-8 | C8H6BrClO | 233.49 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 427890050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5000-66-8
IUPAC Name2-bromo-1-(2-chlorophenyl)ethan-1-one
Molecular FormulaC8H6BrClO
InChI KeyWZWWEVCLPKAQTA-UHFFFAOYSA-N
SMILESClC1=CC=CC=C1C(=O)CBr
さらに表示
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Appearance (Form)Liquid
GC>=94 %
Appearance (Color)Colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only