(R)-(-)-Linalool, 95% (sum of enantiomers)
(R)-(-)-Linalool, 95% (sum of enantiomers)
(R)-(-)-Linalool, 95% (sum of enantiomers)
Thermo Scientific Chemicals

(R)-(-)-Linalool, 95% (sum of enantiomers)

CAS: 126-91-0 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 428311000
Price (USD)
117.65
Special offer
Online exclusive
Ends: 28-Dec-2026
138.00
Save 20.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
117.65
Special offer
Online exclusive
Ends: 28-Dec-2026
138.00
Save 20.35
Each
Chemical Identifiers
CAS126-91-0
IUPAC Name(3R)-3,7-dimethylocta-1,6-dien-3-ol
Molecular FormulaC10H18O
InChI KeyCDOSHBSSFJOMGT-JTQLQIEISA-N
SMILESCC(C)=CCC[C@@](C)(O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4600 to 1.4640 (20°C, 589 nm)
Specific optical rotation-15° to -21° (20°C, 589 nm) (neat)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the enantiomeric purity of (R)-(-)-Linalool, 95% (sum of enantiomers)?

The column used for GC analysis of this (R)-(-)-Linalool, 95% product, yields a single peak for Linalool and cannot differentiate between the two stereoisomers. However, based on the optical rotation value, we are able to declare our product as (R)- Linalool. The value is in accordance with the literature value for the (R)-stereoisomer. Since we do not analyze the counter (S)-stereoisomer in our Linalool product, we cannot verify its presence or absence in our product, nor provide the enantiomeric excess (ee) value of our product.