[4-(1-Piperidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™
[4-(1-Piperidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™
[4-(1-Piperidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™
Thermo Scientific Chemicals

[4-(1-Piperidinylmethyl)phenyl]magnesium bromide, 0.25M solution in THF, AcroSeal™

CAS: 480424-83-7 | C12H16BrMgN | 278.48 g/mol
製品番号(カタログ番号)数量パッケージ
43169050050 mLAcroSeal™ Glass Bottle
製品番号(カタログ番号) 431690500
価格(JPY)
-
数量:
50 mL
パッケージ:
AcroSeal™ Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS480424-83-7
IUPAC Namemagnesium(2+) 4-[(piperidin-1-yl)methyl]benzen-1-ide bromide
Molecular FormulaC12H16BrMgN
InChI KeyXDZALNKSOIYDJE-UHFFFAOYSA-M
SMILES[Mg++].[Br-].C(N1CCCCC1)C1=CC=[C-]C=C1
さらに表示
Appearance (Form)Solution
Appearance (Color)Brown
Titration Grignard0.15 to 0.35 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only