Triethylamine, 99.5%, for analysis
Triethylamine, 99.5%, for analysis
Triethylamine, 99.5%, for analysis
Thermo Scientific Chemicals

Triethylamine, 99.5%, for analysis

CAS: 121-44-8 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 432280025
Price (USD)
267.65
Special offer
Online exclusive
Ends: 28-Dec-2026
315.00
Save 47.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
267.65
Special offer
Online exclusive
Ends: 28-Dec-2026
315.00
Save 47.35
Each
Chemical Identifiers
CAS121-44-8
IUPAC Nametriethylamine
Molecular FormulaC6H15N
InChI KeyZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILESCCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.3990 to 1.4010 (20°C, 589 nm)
GC=<0.05 % (diethylamine)
Residue after evaporation=<0.002 %
Cadmium (Cd)=<0.05 ppm
Chromium (Cr)=<0.02 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only