Ethyl hydrazinoacetate hydrochloride, 97%
Ethyl hydrazinoacetate hydrochloride, 97%
Ethyl hydrazinoacetate hydrochloride, 97%
Thermo Scientific Chemicals

Ethyl hydrazinoacetate hydrochloride, 97%

CAS: 6945-92-2 | C4H11ClN2O2 | 154.59 g/mol
Have Questions?
Catalog NumberQuantityPackaging
43315010010 gGlass bottle
Catalog number 433150100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6945-92-2
IUPAC Namehydrogen ethyl 2-hydrazinylacetate chloride
Molecular FormulaC4H11ClN2O2
InChI KeyHZZRIIPYFPIKHR-UHFFFAOYSA-N
SMILES[H+].[Cl-].CCOC(=O)CNN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or flakes
Infrared spectrumConforms
Melting point146°C to 151°C
Titration Argentometric>=96.0 to =<104.0 %
Appearance (Color)Light yellow to light orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only