4-Bromo-2-chloropyridine, 97%
4-Bromo-2-chloropyridine, 97%
4-Bromo-2-chloropyridine, 97%
Thermo Scientific Chemicals

4-Bromo-2-chloropyridine, 97%

CAS: 73583-37-6 | C5H3BrClN | 192.44 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4348000505 gGlass bottle
Catalog number 434800050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS73583-37-6
IUPAC Name4-bromo-2-chloropyridine
Molecular FormulaC5H3BrClN
InChI KeyONHMWUXYIFULDO-UHFFFAOYSA-N
SMILESClC1=CC(Br)=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to white to yellow to orange
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only