Iron(III) nitrate nonahydrate, 99.9995%, (trace metal basis)
Iron(III) nitrate nonahydrate, 99.9995%, (trace metal basis)
Iron(III) nitrate nonahydrate, 99.9995%, (trace metal basis)
Thermo Scientific Chemicals

Iron(III) nitrate nonahydrate, 99.9995%, (trace metal basis)

CAS: 7782-61-8 | FeH18N3O18 | 403.99 g/mol
Catalog NumberQuantityPackaging
43681025025 gGlass bottle
Catalog number 436810250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7782-61-8
IUPAC Nameiron(3+) nonahydrate trinitrate
Molecular FormulaFeH18N3O18
InChI KeySZQUEWJRBJDHSM-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Iodometric13.5 to 14.2 % (Fe)
Appearance (Form)Crystals
Total trace metal impurities=<5 ppm
Appearance (Color)Off-white to violet to purple
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only