Zinc sulfate heptahydrate, granular, USP, FCC, GMP, J.T.Baker™
Avantor™

Zinc sulfate heptahydrate, granular, USP, FCC, GMP, J.T.Baker™

Food GMP Manufactured Product and meets USP and F.C.C Requirements.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
12 kg
Katalognummer 4384-01
Preis (EUR)
-
Menge:
500 g
Food GMP Manufactured Product and meets USP and F.C.C Requirements.
Specifications
CAS7446-20-0
Chemischer Name oder MaterialZinc sulfate heptahydrate
FarbeWeiß
Schmelzpunkt100°C
SummenformelZnSO4·7H2O
VerpackungWhite poly bottle, tamper-evident cap
SynonymZinc Sulfate hydrate, Zinc sulphate heptahydrate, ZnSO4
GüteUSP
Molekulargewicht (g/mol)287.54
Reinheit (%)99.0-108.7%
Physikalische FormCrystalline Powder
Menge500 g
Unit SizeEach