N-Hydroxysulfosuccinimide sodium salt, 95%
N-Hydroxysulfosuccinimide sodium salt, 95%
N-Hydroxysulfosuccinimide sodium salt, 95%
Thermo Scientific Chemicals

N-Hydroxysulfosuccinimide sodium salt, 95%

Coupling reagent | CAS: 106627-54-7 | C4H4NNaO6S | 217.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 438650050
Price (EUR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS106627-54-7
IUPAC Namesodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
Molecular FormulaC4H4NNaO6S
InChI KeyRPENMORRBUTCPR-UHFFFAOYNA-M
SMILES[Na+].ON1C(=O)CC(C1=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagent
RUO – Research Use Only