FMOC-O-tert-Butyl-L-tyrosine, 98%
FMOC-O-tert-Butyl-L-tyrosine, 98%
FMOC-O-tert-Butyl-L-tyrosine, 98%
Thermo Scientific Chemicals

FMOC-O-tert-Butyl-L-tyrosine, 98%

CAS: 71989-38-3 | C28H29NO5 | 459.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Catalog number 439090100
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.90
Save 6.25
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
38.65
Special offer
Online exclusive
Ends: 28-Dec-2026
44.90
Save 6.25
Each
Chemical Identifiers
CAS71989-38-3
IUPAC Name(2S)-3-[4-(tert-butoxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Molecular FormulaC28H29NO5
InChI KeyJAUKCFULLJFBFN-KSYWNVGFNA-N
SMILESCC(C)(C)OC1=CC=C(C[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
Specific optical rotation-27° to -31° (20°C, 589 nm) (c=1 in DMF)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only