Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Complex III Antibodies

          Invitrogen

          Complex III Monoclonal Antibody (11A51H12)

          View all (3) Complex III antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Complex III Monoclonal Antibody (11A51H12)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Complex III Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.
          Complex III Antibody in Immunoprecipitation (IP)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Complex III Antibody (43-9400) in IP

          Immunoprecipitation analysis of UQCRC1 using a UQCRC1 Monoclonal antibody (Product # 43-9400) at 10 µg/mg of lysate. Lane 1: 0.1 mg of Bovine mitochondria. Lane 2: 0.05 mg Bovine mitochondria. Lane 3: 0.05 mg Human mitochondria. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Complex III Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          43-9400

          Applications
          Tested Dilution
          Publications

          Immunoprecipitation (IP)

          100 µg/mg
          -
          Product Specifications

          Species Reactivity

          Bovine, Human

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          11A51H12

          Immunogen

          Bovine Heart Complex III

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Agarose
          • Request custom conjugation

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          HEPES buffered saline

          Contains

          0.02% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533546

          Product Specific Information

          Positive control: Bovine heart and Human mitochondria

          Target Information

          Complex III, or ubiquinol cytochrome c oxidoreductase, is a complex of 11 subunits and catalyses electron transfer from reduced ubiquinone or coenzyme Q 10 to cytochrome c. The complex contains 11 different subunits, one of which, cytochrome b, is encoded on mitochondrial DNA, while the remainder subunits are nuclear-encoded. Genetic complex III deficiencies occur but are rare.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: complex III subunit 1; complex III subunit 10; complex III subunit 2; complex III subunit 4; complex III subunit 5; complex III subunit 6; complex III subunit 7; complex III subunit 8; Complex III subunit 9; complex III subunit IV; complex III subunit VII; complex III subunit VIII; complex III subunit X; complex III subunit XI; core protein I; core protein II; core protein II precursor; CYC1; cytochrome b-c1 complex subunit 4; cytochrome b-c1 complex subunit 5; cytochrome bc-1 complex core protein II; cytochrome c-1; cytochrome c1; cytochrome c1 non-heme 11 kDa protein; cytochrome c1 non-heme 7 kDa protein; cytochrome C1, nonheme 7kDa protein; epididymis secretory sperm binding protein; low molecular mass ubiquinone-binding protein (9.5kD); mitochondrial hinge protein; mitochondrial hinge protein precursor; mitochondrial ubiquinone-binding protein; QP-C; Rieske iron-sulfur protein; Rieske iron-sulfur protein precursor; Rieske protein UQCRFS1; Subunit 9; ubiquinol-cytochrome c reductase (6.4kD) subunit; ubiquinol-cytochrome c reductase complex (7.2 kD); ubiquinol-cytochrome c reductase complex 11 kDa protein; ubiquinol-cytochrome c reductase complex 14 kDa protein; ubiquinol-cytochrome c reductase complex 6.4 kDa protein; ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase core protein I; ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, 6.4kDa subunit; ubiquinol-cytochrome c reductase, complex III subunit VI; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; ubiquinol-cytochrome c reductase, complex III subunit VIII; ubiquinol-cytochrome c reductase, complex III subunit X, 7.2kDa; ubiquinol-cytochrome-c reductase complex core protein 1; ubiquinol-cytochrome-c reductase complex core protein 2; ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; ubiquinone-binding protein; ubiquinone-binding protein precursor; UCRC; unnamed protein product; UQCRB

          View more View less

          Gene Aliases: 0710008D09Rik; D3S3191; HSPC051; HSPC119; HSPC151; ISP; MC3DN10; MC3DN11; MC3DN3; MC3DN4; MC3DN5; MC3DN6; PKNPY; QCR1; QCR10; QCR2; QCR6; QCR7; QCR8; QCR9; QP-C; QPC; RIP1; RIS1; RISP; UCCR7.2; UCRC; UQBC; UQBP; UQCR; UQCR1; UQCR2; UQCR4; UQCR5; UQCR6; UQCR7; UQCR8; UQPC

          View more View less

          UniProt ID: (Bovine) P00125, (Human) P08574, (Bovine) P00130, (Human) Q9UDW1, (Human) O14957, (Bovine) P07552, (Bovine) P00129, (Human) P14927, (Bovine) P31800, (Human) P31930, (Bovine) P23004, (Human) P22695, (Bovine) P13272, (Human) P47985, (Bovine) P00126, (Human) P07919, (Human) O14949, (Bovine) P13271

          View more View less

          Entrez Gene ID: (Bovine) 512500, (Human) 1537, (Bovine) 616109, (Human) 29796, (Human) 10975, (Bovine) 281570, (Bovine) 616871, (Human) 7381, (Bovine) 282393, (Human) 7384, (Bovine) 282394, (Human) 7385, (Bovine) 287020, (Human) 7386, (Bovine) 613899, (Human) 7388, (Human) 27089, (Bovine) 286885

          View more View less

          Function(s)
          heme binding metal ion binding protein binding ubiquinol-cytochrome-c reductase activity electron carrier activity catalytic activity ubiquitin protein ligase binding macromolecular complex binding metalloendopeptidase activity 2 iron, 2 sulfur cluster binding oxidoreductase activity metalloprotease reductase oxidoreductase metabolite interconversion enzyme
          Process(es)
          mitochondrial electron transport, ubiquinol to cytochrome c response to glucagon cellular respiration hydrogen ion transmembrane transport generation of precursor metabolites and energy electron transport chain oxidative phosphorylation aerobic respiration oxidation-reduction process response to activity response to alkaloid protein localization to mitochondrion proteolysis respiratory electron transport chain mitochondrial respiratory chain complex III assembly subthalamus development pons development cerebellar Purkinje cell layer development hippocampus development thalamus development hypothalamus development pyramidal neuron development midbrain development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-76c4fffd86-nbmw6:80/100.66.77.38:80.
          git-commit: bdc067a1e9e92d056899a2dda64af631988aa8a1
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.54.0-Offline