1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%
Acros Organics

1-Isopropyl-1H-pyrazole-4-boronic acid pinacol ester, 97%

CAS: 879487-10-2 | C12H21BN2O2 | 236.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 445090010
also known as 44509
HSN 29331990
Price (INR)
38,979.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
38,979.00
Each
Chemical Identifiers
CAS879487-10-2
IUPAC Name1-(propan-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular FormulaC12H21BN2O2
InChI KeyOGYYMVGDKVJYSU-UHFFFAOYSA-N
SMILESCC(C)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to light brown
Appearance (Form)Solid
Infrared spectrumConforms
Melting point38°C to 46°C
GC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only