Citric acid, trisodium salt dihydrate, ACS reagent
Citric acid, trisodium salt dihydrate, ACS reagent
Citric acid, trisodium salt dihydrate, ACS reagent
Citric acid, trisodium salt dihydrate, ACS reagent
Thermo Scientific Chemicals

Citric acid, trisodium salt dihydrate, ACS reagent

CAS: 6132-04-3 | C6H5Na3O7·2H2O | 294.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 446330050
Price (USD)
205.65
Online Exclusive
228.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
205.65
Online Exclusive
228.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS6132-04-3
IUPAC Nametrisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
Molecular FormulaC6H9Na3O9
InChI KeyNLJMYIDDQXHKNR-UHFFFAOYSA-K
SMILESO.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Chloride (Cl)=<0.003 %
Ammonia (NH3)=<0.003 %
Iron (Fe)=<5 ppm
Appearance (Form)Crystalline powder or crystals
Titration with HClO4>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only