Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Sodium bicarbonate, 99.5%, for biochemistry
Thermo Scientific Chemicals

Sodium bicarbonate, 99.5%, for biochemistry

CAS: 144-55-8 ∣ CHNaO3 ∣ 84.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4471000101 kgPlastic Bottle
Catalog number 447100010
Price (USD)
134.00
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
134.00
Each
Chemical Identifiers
CAS144-55-8
IUPAC Namesodium hydrogen carbonate
Molecular FormulaCHNaO3
InChI KeyUIIMBOGNXHQVGW-UHFFFAOYSA-M
SMILES[Na+].OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with HCl99.4 to 100.6 %
SolubilityClear colorless
Heavy metals (as Pb)=<5 ppm
UVat 280 nm A: =<0.02
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only