Mercuric acetate, ACS reagent
Mercuric acetate, ACS reagent
Mercuric acetate, ACS reagent
Thermo Scientific Chemicals

Mercuric acetate, ACS reagent

CAS: 1600-27-7 | C4H6HgO4 | 318.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 447275000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1600-27-7
IUPAC Namemercury(2+) diacetate
Molecular FormulaC4H6HgO4
InChI KeyBRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES[Hg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Titration with NH4SCN>=98.0 %
Insoluble matter=<0.01 %
Chloride (Cl)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only