Lithium sulfate, for analysis, anhydrous
Lithium sulfate, for analysis, anhydrous
Lithium sulfate, for analysis, anhydrous
Thermo Scientific Chemicals

Lithium sulfate, for analysis, anhydrous

CAS: 10377-48-7 | Li2O4S | 109.94 g/mol
Quantity:
250 g
1 kg
Catalog number 447550010
Price (JPY)
-
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS10377-48-7
IUPAC Namedilithium(1+) sulfate
Molecular FormulaLi2O4S
InChI KeyINHCSSUBVCNVSK-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Potassium (K)=<0.02 %
Iron (Fe)=<0.005 %
Magnesium (Mg)=<0.02 %
Appearance (Form)Crystalline powder
Water=<1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only