Sodium phosphate, dibasic dodecahydrate, 99%, extra pure
Sodium phosphate, dibasic dodecahydrate, 99%, extra pure
Sodium phosphate, dibasic dodecahydrate, 99%, extra pure
Thermo Scientific Chemicals

Sodium phosphate, dibasic dodecahydrate, 99%, extra pure

CAS: 10039-32-4 ∣ HNa2O4P·12H2O ∣ 358.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 448160010
Price (USD)
70.10
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
70.10
Each
Chemical Identifiers
CAS10039-32-4
IUPAC Namedisodium dodecahydrate hydrogen phosphate
Molecular FormulaH25Na2O16P
InChI KeyDGLRDKLJZLEJCY-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystals
Water57 to 61 %
Titration with HClO4>=98.5 %
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only