3-Amino-1-bromoisoquinoline, 97%
3-Amino-1-bromoisoquinoline, 97%
3-Amino-1-bromoisoquinoline, 97%
Thermo Scientific Chemicals

3-Amino-1-bromoisoquinoline, 97%

CAS: 13130-79-5 | C9H7BrN2 | 223.07 g/mol
Have Questions?
Catalog NumberQuantity
4491600101 g
Catalog number 449160010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS13130-79-5
IUPAC Name1-bromoisoquinolin-3-amine
Molecular FormulaC9H7BrN2
InChI KeyPSQUIUNIVDKHJK-UHFFFAOYSA-N
SMILESNC1=NC(Br)=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point143°C to 147°C
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Yellow to light brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only