(S)-Styrene oxide, 94%
(S)-Styrene oxide, 94%
(S)-Styrene oxide, 94%
Thermo Scientific Chemicals

(S)-Styrene oxide, 94%

CAS: 20780-54-5 | C8H8O | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 449450050
Price (USD)
139.65
Special offer
Online exclusive
Ends: 28-Dec-2026
164.00
Save 24.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
139.65
Special offer
Online exclusive
Ends: 28-Dec-2026
164.00
Save 24.35
Each
Chemical Identifiers
CAS20780-54-5
IUPAC Name(2S)-2-phenyloxirane
Molecular FormulaC8H8O
InChI KeyAWMVMTVKBNGEAK-MRVPVSSYSA-N
SMILESC1O[C@H]1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=92 %
Appearance (Color)Colorless to light yellow
Specific optical rotation-31° to -35° (20°C, 589 nm) (neat)
Refractive index1.5340 to 1.5360 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only