Thermo Scientific Chemicals

Atenolol, 98%, Thermo Scientific Chemicals

Catalog number: 449550250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Atenolol, 98%, Thermo Scientific Chemicals

Catalog number: 449550250
25 g, Glass bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
29122-68-7
IUPAC Name
2-(4-{2-hydroxy-3-[(propan-2-yl)amino]propoxy}phenyl)acetamide
Molecular Formula
C14H22N2O3
InChI Key
METKIMKYRPQLGS-UHFFFAOYNA-N
SMILES
CC(C)NCC(O)COC1=CC=C(CC(N)=O)C=C1
Infrared spectrum
Conforms
Appearance (Color)
White to off-white
Melting point
151°C to 155°C
Appearance (Form)
Powder
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text