Rosuvastatin, calcium salt, 98%
Rosuvastatin, calcium salt, 98%
Rosuvastatin, calcium salt, 98%
Thermo Scientific Chemicals

Rosuvastatin, calcium salt, 98%

CAS: 147098-20-2 | C22H28FN3O61/2Ca | 500.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 456232500
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
161.00
Save 24.35
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
136.65
Special offer
Online exclusive
Ends: 28-Dec-2026
161.00
Save 24.35
Each
Chemical Identifiers
CAS147098-20-2
IUPAC Namecalcium bis((3R,5S,6E)-7-[4-(4-fluorophenyl)-2-(N-methylmethanesulfonamido)-6-(propan-2-yl)pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate)
Molecular FormulaC44H54CaF2N6O12S2
InChI KeyLALFOYNTGMUKGG-BGRFNVSISA-L
SMILES[Ca++].CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1\C=C\[C@@H](O)C[C@@H](O)CC([O-])=O)N(C)S(C)(=O)=O.CC(C)C1=NC(=NC(C2=CC=C(F)C=C2)=C1\C=C\[C@@H](O)C[C@@H](O)CC([O-])=O)N(C)S(C)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only