Furfuryl isocyanate, 97%
Furfuryl isocyanate, 97%
Furfuryl isocyanate, 97%
Thermo Scientific Chemicals

Furfuryl isocyanate, 97%

CAS: 71189-15-6 | C6H5NO2 | 123.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 456490050
Price (USD)
170.65
Special offer
Online exclusive
Ends: 28-Dec-2026
201.00
Save 30.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
170.65
Special offer
Online exclusive
Ends: 28-Dec-2026
201.00
Save 30.35
Each
Chemical Identifiers
CAS71189-15-6
IUPAC Name2-(isocyanatomethyl)furan
Molecular FormulaC6H5NO2
InChI KeyUIADMYLYGJYUSQ-UHFFFAOYSA-N
SMILESO=C=NCC1=CC=CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4750 to 1.4770 (20°C, 589 nm)
Appearance (Color)Clear colorless to brown
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only