L-Lysine methyl ester dihydrochloride, 98%
L-Lysine methyl ester dihydrochloride, 98%
L-Lysine methyl ester dihydrochloride, 98%
Thermo Scientific Chemicals

L-Lysine methyl ester dihydrochloride, 98%

CAS: 26348-70-9 | C7H18Cl2N2O2 | 233.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 459271000
Price (USD)
229.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
229.00
Each
Chemical Identifiers
CAS26348-70-9
IUPAC Namedihydrogen methyl (2S)-2,6-diaminohexanoate dichloride
Molecular FormulaC7H18Cl2N2O2
InChI KeySXZCBVCQHOJXDR-ILKKLZGPSA-N
SMILES[H+].[H+].[Cl-].[Cl-].COC(=O)[C@@H](N)CCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Specific optical rotation+21° to +25° (20°C, 589 nm) (1% in MeOH)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only