Cyproterone acetate, 98%, Thermo Scientific Chemicals
Cyproterone acetate, 98%, Thermo Scientific Chemicals
Cyproterone acetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyproterone acetate, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
459542500250mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 459542500
Price (USD)
-
Quantity:
250mg
Request bulk or custom format
Chemical Identifiers
CAS427-51-0
IUPAC Name(1S,2S,3S,5R,11R,12S,15R,16S)-15-acetyl-9-chloro-2,16-dimethyl-6-oxopentacyclo[9.7.0.0²,⁸.0³,⁵.0¹²,¹⁶]octadeca-7,9-dien-15-yl acetate
Molecular FormulaC24H29ClO4
InChI KeyUWFYSQMTEOIJJG-FDTZYFLXSA-N
SMILESCC(=O)O[C@@]1(CC[C@H]2[C@@H]3C=C(Cl)C4=CC(=O)[C@@H]5C[C@@H]5[C@]4(C)[C@H]3CC[C@]12C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point208°C to 215°C
HPLC>=97.5 %
Loss on drying=<0.5 % (80°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only