Hamburger Menu Button
Thermo Fisher Scientific Logo
๋กœ๊ทธ์ธ
ํšŒ์›์ด ์•„๋‹ˆ์‹ ๊ฐ€์š”? ๊ณ„์ • ์ƒ์„ฑํ•˜๊ธฐ
  • ๋ชจ๋“  ์ œํ’ˆ ์ฃผ๋ฌธ
    • ํ•ญ์ฒด
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR ๋ถ„์„
    • ํ”ผํŽซ ๋ฐ ํ”ผํŽซ ํŒ
    • ์‹คํ—˜์‹ค์šฉ ์›์‹ฌ๋ถ„๋ฆฌ๊ธฐ
    • ์ดˆ์ €์˜จ ๋ƒ‰๋™๊ณ 
    • ๋ถ„๊ด‘ํ•™
    • ๋น„์ด์ปค
    • PCR ์žฅ๋น„ ๋ฐ ์šฉํ’ˆ
    • ์ œํ’ˆ ์นดํ…Œ๊ณ ๋ฆฌ ์ „์ฒด๋ณด๊ธฐ
  • ์‘์šฉ ๋ถ„์•ผ ๋ฐ ๊ธฐ๋ฒ•
    • ์„ธํฌ ๋ถ„์„
    • ์‹คํ—˜์‹ค ์žฅ๋น„
    • Real-Time PCR
    • PCR
    • ํฌ๋กœ๋งˆํ† ๊ทธ๋ž˜ํ”ผ
    • ์„ธํฌ ๋ฐฐ์–‘ ๋ฐ ํŠธ๋žœ์ŠคํŽ™์…˜
    • DNA/RNA ์ถ”์ถœ๊ณผ ๋ถ„์„
    • ๋‹จ๋ฐฑ์งˆ ์ƒ๋ฌผํ•™
    • ์œ ์„ธํฌ๋ถ„์„
    • ํ™”ํ•™ ์ œํ’ˆ
    • ์‘์šฉ ๋ถ„์•ผ ๋ฐ ๊ธฐ๋ฒ• ์ „์ฒด๋ณด๊ธฐ
  • ์„œ๋น„์Šค
    • ๋งž์ถคํ˜• ์„œ๋น„์Šค
    • ์—”ํ„ฐํ”„๋ผ์ด์ฆˆ๊ธ‰ ์‹คํ—˜์‹ค ์ •๋ณด ์ฒ˜๋ฆฌ
    • 360ยฐ CDMO ๋ฐ CRO ์„œ๋น„์Šค
    • CDMO ์„œ๋น„์Šค
    • ์ž„์ƒ์‹œํ—˜ CRO ์„œ๋น„์Šค
    • ์žฅ๋น„ ์„œ๋น„์Šค
    • ๊ต์œก ์„œ๋น„์Šค
    • Unity Lab Services
    • ์„œ๋น„์Šค ์ „์ฒด ๋ณด๊ธฐ
  • ์ง€์›
    • ๊ณ ๊ฐ์„ผํ„ฐ
    • ๋ฌธ์˜ํ•˜๊ธฐ
    • ์‹œํ—˜์„ฑ์ ์„œ(COA) ๋ฐ ์ ํ•ฉ์„ฑ ์ธ์ฆ์„œ(COC)
    • Safety Data Sheets (SDS)
    • ๋งค๋‰ด์–ผ
    • ์ธ์šฉ ๋ฐ ์ฐธ๊ณ  ๋ฌธํ—Œ
    • ๊ธฐ๊ธฐ ์ง€์›
    • ๊ธฐ์ˆ  ์ž๋ฃŒ/์ œํ’ˆ FAQ
    • ํ•™์Šต ์„ผํ„ฐ
    • ์ง€์› ๋ฉ”๋‰ด ์ „์ฒด๋ณด๊ธฐ
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • ์‹ ์ œํ’ˆ
  • ๋ฌธ์˜ํ•˜๊ธฐ
  • ๋น ๋ฅธ ์ฃผ๋ฌธ
  • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
  • ์ œํ’ˆ ๋ฌธ์„œ ๊ฒ€์ƒ‰
Thermo Fisher Scientific Logo

Search

์ „์ฒด๊ฒ€์ƒ‰
Search button
          • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
          • ๋น ๋ฅธ์ฃผ๋ฌธ
          • ๋กœ๊ทธ์ธ
            ๋กœ๊ทธ์ธ
            ํšŒ์›์ด ์•„๋‹ˆ์‹ ๊ฐ€์š”? ๊ณ„์ • ์ƒ์„ฑํ•˜๊ธฐ
            • ๊ณ„์ •์ •๋ณด
            • ์ฃผ๋ฌธ๋‚ด์—ญ์กฐํšŒ
            • ์ปค์Šคํ…€์ œํ’ˆ ๋ฐ ํ”„๋กœ์ ํŠธ
            • Services Central
          • Primary Antibodies ›
          • CSF3R (CD114) Antibodies

          Invitrogen

          CD114 (G-CSFR) Monoclonal Antibody (LMM741), PerCP-eFluor™ 710, eBioscience™

          View all (25) CSF3R (CD114) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite CD114 (G-CSFR) Monoclonal Antibody (LMM741), PerCP-eFluor™ 710, eBioscience™

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          CD114 (G-CSFR) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD114 (G-CSFR) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD114 (G-CSFR) Antibody (46-1149-42) in Flow

          Staining of normal human peripheral blood cells with Anti-Human CD114 (G-CSFR) PerCP-eFluorยฎ 710. Cells in the lymphocyte (blue histogram), monocyte (orange histogram), and granulocyte (purple histogram) gates were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD114 (G-CSFR) Antibody in Flow Cytometry (Flow)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as โ€œraw-uneditedโ€, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          46-1149-42

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          5 ยตL (0.25 ยตg)/test
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1, kappa

          Recommended Isotype Control

          Mouse IgG1 kappa Isotype Control (P3.6.2.8.1), PerCP-eFluorโ„ข 710, eBioscienceโ„ข

          Class

          Monoclonal

          Type

          Antibody

          Clone

          LMM741

          Conjugate

          PerCP-eFluorโ„ข 710 PerCP-eFluorโ„ข 710 PerCP-eFluorโ„ข 710

          Excitation/Emission Max

          482/708 nm View spectra spectra

          Form

          Liquid

          Concentration

          5 ยตL/Test

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4ยฐC, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_11045508

          Product Specific Information

          Description: This LMM741 monoclonal antibody reacts with human CD114, which is also known as granulocyte colony-stimulating factor receptor (G-CSFR). A type I hematopoietin cytokine receptor, CD114 is expressed on granulocytes such as neutrophils, myeloid cells, monocytes, and leukemic cells. This receptor is absent from lymphocytes, erythrocytes, and eosinophils. CD114 can also be detected on non-hematopoietic cells, including neurons, endothelial cells, and cardiomyocytes. Studies demonstrate that expression of CD114 can be downregulated by LPS, TNF, phorbol ester, and G-CSF itself. Binding of G-CSF binding induces homodimerization of the receptor that activates signaling transduction (e.g., the JAK/STAT and Erk1/2 pathways) to mediate cell proliferation, survival, and differentiation.

          Applications Reported: This LMM741 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This LMM741 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 ยตL (0.25 ยตg) per test. A test is defined as the amount (ยตg) of antibody that will stain a cell sample in a final volume of 100 ยตL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

          PerCP-eFluorยฎ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.

          Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-822-49) (100 ยตL cell sample + 100 ยตL IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333-54) for up to 3 days in the dark at 4ยฐC with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.

          Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.

          Filtration: 0.2 ยตm post-manufacturing filtered.

          Target Information

          CSF3R is the receptor for colony stimulating factor 3, a cytokine that controls the production, differentiation, and function of granulocytes. The encoded protein, which is a member of the family of cytokine receptors, may also function in some cell surface adhesion or recognition processes. Four transcript variants encoding four different isoforms have been found for this gene, with three of the isoforms being membrane-bound and the other being secreted and soluble.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CD114 antigen; colony stimulating factor 3 receptor (granulocyte); CSF3R; G-CSF receptor; G-CSF-R

          View more View less

          Gene Aliases: CD114; GCSFR; SCN7

          View more View less

          UniProt ID: (Human) Q99062

          View more View less

          Entrez Gene ID: (Human) 1441

          View more View less

          Function(s)
          cytokine receptor activity protein binding cytokine binding signaling receptor activity granulocyte colony-stimulating factor binding
          Process(es)
          defense response signal transduction positive regulation of cell proliferation cytokine-mediated signaling pathway amelogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          ์˜จ๋ผ์ธ ์ฃผ๋ฌธ Plus Icon Minus Icon
          • ์ฃผ๋ฌธ ํ˜„ํ™ฉ
          • ์ฃผ๋ฌธ ์ง€์›
          • ๋น ๋ฅธ ์ฃผ๋ฌธ
          • Supply Center
          • eProcurement
          ์ง€์› Plus Icon Minus Icon
          • Help and Support
          • ๊ณ ๊ฐ ์„ผํ„ฐ
          • ๊ธฐ์ˆ  ์ง€์› ์„ผํ„ฐ
          • ์ œํ’ˆ ๋ฌธ์„œ ๊ฒ€์ƒ‰
          • ์‚ฌ์ดํŠธ ๋ฌธ์ œ ๋ณด๊ณ 
          ๊ต์œก ๋ฐ ์ด๋ฒคํŠธ Plus Icon Minus Icon
          • ๊ต์œก ์„ผํ„ฐ
          • ํ”„๋กœ๋ชจ์…˜
          • ์ด๋ฒคํŠธ ๋ฐ ์›จ๋น„๋‚˜
          • ์†Œ์…œ ๋ฏธ๋””์–ด
          About Thermo Fisher Plus Icon Minus Icon
          • ์†Œ๊ฐœ ์†Œ๊ฐœ
          • ์ฑ„์šฉ ์ฑ„์šฉ
          • ํˆฌ์ž์ž ํˆฌ์ž์ž
          • ๋‰ด์Šค ๋‰ด์Šค
          • ์‚ฌํšŒ์  ์ฑ…์ž„ ์‚ฌํšŒ์  ์ฑ…์ž„
          • Trademarks
          • ๊ณต์ •๊ฑฐ๋ž˜ ๊ณต์ •๊ฑฐ๋ž˜
          • ์ •์ฑ… ๋ฐ ๊ณ ์ง€
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • ์ด์šฉ ์•ฝ๊ด€
          • ๊ฐœ์ธ์ •๋ณด์ฒ˜๋ฆฌ๋ฐฉ์นจ
          • ๊ฐ€๊ฒฉ ๋ฐ ์šด์ž„ ์ •์ฑ…
          ยฉ 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด

           

          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat

          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด

           

          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat

          ๊ณ ๊ฐ์„ผํ„ฐ ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-9555   |   Live Chat   |   ์นด์นด์˜คํ†ก ์ƒ๋‹ด

           

          ์žฅ๋น„ ์„œ๋น„์Šค ๋ฌธ์˜ | ํ‰์ผ 09:00~18:00
          1661-5055   |   Live Chat

          ์จ๋ชจ ํ”ผ์…” ์‚ฌ์ด์–ธํ‹ฐํ”ฝ ์ฝ”๋ฆฌ์•„ ์ฃผ์‹ํšŒ์‚ฌ
          ๋Œ€ํ‘œ์ž : ์„์ˆ˜์ง„
          ์‚ฌ์—…์ž ๋“ฑ๋ก๋ฒˆํ˜ธ : 117-81-46910
          ์ž…๊ธˆ๊ณ„์ขŒ : ํ•˜๋‚˜์€ํ–‰ 336-890014-06204
          (์˜ˆ๊ธˆ์ฃผ : ์จ๋ชจํ”ผ์…”์‚ฌ์ด์–ธํ‹ฐํ”ฝ์ฝ”๋ฆฌ์•„ ์ฃผ์‹ํšŒ์‚ฌ)

           

          ์จ๋ชจ ํ”ผ์…” ์‚ฌ์ด์–ธํ‹ฐํ”ฝ ์†”๋ฃจ์…˜์Šค ์œ ํ•œํšŒ์‚ฌ
          ๋Œ€ํ‘œ์ž : ์„์ˆ˜์ง„
          ์‚ฌ์—…์ž ๋“ฑ๋ก๋ฒˆํ˜ธ : 114-86-04783
          ์ž…๊ธˆ๊ณ„์ขŒ : ์‹ ํ•œ์€ํ–‰ 140-004-396660
          (์˜ˆ๊ธˆ์ฃผ : ์จ๋ชจํ”ผ์…”์‚ฌ์ด์–ธํ‹ฐํ”ฝ์†”๋ฃจ์…˜์Šค ์œ ํ•œํšŒ์‚ฌ)

           

          ์ฃผ์†Œ : ์„œ์šธ์‹œ ๊ฐ•๋‚จ๊ตฌ ๊ด‘ํ‰๋กœ 281 ์ˆ˜์„œ์˜คํ”ผ์Šค๋นŒ๋”ฉ 12์ธต 06349 | ํ†ต์‹ ํŒ๋งค์—…์‹ ๊ณ ๋ฒˆํ˜ธ : 2015-์„œ์šธ๊ฐ•๋‚จ-00898

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-p4d9w:80/100.66.75.182:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0