Ifosfamide
Ifosfamide
Ifosfamide
Thermo Scientific Chemicals

Ifosfamide

CAS: 3778-73-2 | C7H15Cl2N2O2P | 261.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 461350010
Price (EUR)
149,00
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
149,00
Each
Chemical Identifiers
CAS3778-73-2
IUPAC Name3-(2-chloroethyl)-2-[(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one
Molecular FormulaC7H15Cl2N2O2P
InChI KeyHOMGKSMUEGBAAB-UHFFFAOYNA-N
SMILESClCCNP1(=O)OCCCN1CCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.5 %
Water=<0.5 %
Appearance (Color)White to almost white to light beige
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only