Product Image
Thermo Scientific Chemicals

Metoclopramide hydrochloride, 98%

CAS: 7232-21-5 | C14H23Cl2N3O2 | 336.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 461820050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS7232-21-5
IUPAC Name4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrochloride
Molecular FormulaC14H23Cl2N3O2
InChI KeyRVFUNJWWXKCWNS-UHFFFAOYSA-N
SMILESCl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Appearance (Color)White to off-white
Water=<7.5 %
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only