Axiom™ Caprine Genotyping v2 Array, 384HT format
Applied Biosystems™
Axiom™ Caprine Genotyping v2 Array, 384HT format
The Axiom Caprine Genotyping v2 Array is a component of our Axiom Agrigenomics Genotyping solution where target genomic DNA is詳細を見る
製品番号(カタログ番号)数量
5513431 array
製品番号(カタログ番号) 551343
価格(JPY)
-
数量:
1 array
The Axiom Caprine Genotyping v2 Array is a component of our Axiom Agrigenomics Genotyping solution where target genomic DNA is prepared and processed on the GeneTitan Multi-Channel instrument. The automated process includes hybridization of gDNA to this high-throughput microarray followed by staining, washing, imaging, and output of genotyping data for further analysis.
The Axiom Caprine Genotyping v2 Array was designed in collaboration with the International Goat Genome Consortium and the VarGoats Consortium. It includes single-nucleotide polymorphisms (SNPs) from the Illumina Goat_IGGC_65K_v2 and IMAGE 10K goat chip [more info (PDF)]. This array also includes insertions, deletions, and duplications targeted by 1 to 50 probes along with 59,795 SNPs and 265 structural variants.
This array is also available in mini-96 format.
Features of the Axiom Caprine Genotyping v2 Array include:
• An updated design that includes the original goat SNP panel, allowing compatibility with previous studies
• An innovative design that is the first attempt to target structural variants (this array includes 265 structural variants)
Array content
• Contains 59,795 SNPs selected for high-throughput production arrays based on genome coverage and best compromise to include the largest number of informative SNPs for goat species
• The following SNPs were included in the design of this array: SNPs with different MAF profiles, SNPs chosen in breeds not represented in the discovery dataset, SNPs that are able to join current GBS data and fill the remaining gaps including mitochondrial markers, SNPs in specific regions, Y markers, and structural variants
Applications
• Genome-wide association studies
• Molecular breeding: association mapping and genomic selection
• Creation of custom array by selecting SNPs from this array (every SNP chosen has been demonstrated to perform well)
Required products
GeneChip Command Console Software
GeneTitan Multi-Channel Instrument
Axiom Analysis Suite Software
Axiom Long Format Export Tool (AxLE)
研究用途にのみ使用できます
仕様
使用対象 (装置)Axiom Reagents
製品ラインAxiom
数量1 array
出荷条件Room temperature
アレイの数384 arrays
種Caprine
Unit Size1 array