Search
Search
FIGURE: 1 / 5
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: The UCHL1 monoclonal antibody reacts with human CD45RO, a 180 kDa isoform of CD45. CD45RO is expressed by most thymocytes, activated memory T cells, granulocytes and monocytes. CD22 is a ligand for CD45RO. Expression of CD45RO and CD45RA is used commonly to discriminate subsets of peripheral T cells.
Applications Reported: The UCHL1 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.
Applications Tested: The UCHL1 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue and can be used at 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
Using conjugate solutions: Centrifuge the protein conjugate solution briefly in a microcentrifuge before use; add only the supernatant to the experiment. This step will help eliminate any protein aggregates that may have formed during storage, thereby reducing nonspecific background staining.
CD45RO (UCHL1, ubiquitin carboxyl terminal esterase L1) is the shortest isoform of a receptor-type protein tyrosine phosphatase, CD45 glycoprotein. CD45 is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases, promotes cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 isoforms differ in their extracellular domains, whereas they share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45RO is expressed on macrophages, CD8+ T cells, activated T cells and myeloma cells. Besides the role in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD45 antigen; glycoprotein 180; protein tyrosine phosphatase; protein tyrosine phosphatase, receptor type, c polypeptide; T200 glycoprotein; T200 leukocyte common antigen; unnamed protein product
Gene Aliases: B220; CD45; CD45R; GP180; IMD105; L-CA; LCA; LY5; T200
UniProt ID: (Human) P08575
Entrez Gene ID: (Human) 5788
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support