Sodium hydroxide, 95.0-100.5%, pellets, NF, FCC, GMP, Macron Fine Chemicals™
Avantor™

Sodium hydroxide, 95.0-100.5%, pellets, NF, FCC, GMP, Macron Fine Chemicals™

This product is a GMP-manufactured chemical that meets both the F.C.C. and N.F. Requirements.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 g
2.5 kg
12 kg
25 kg
110 lb.
Verpackung:
Alle anzeigen
Mall stakmor case
Polyethylene pail, screw top
Katalognummer 7680-20
Preis (EUR)
-
Menge:
12 kg
Verpackung:
Mall stakmor case
This product is a GMP-manufactured chemical that meets both the F.C.C. and N.F. Requirements.
This product is a GMP-manufactured chemical that meets both the F.C.C. and N.F. Requirements.
Specifications
Siedepunkt1390°C (1013 hPa)
CAS1310-73-2
Chemischer Name oder MaterialSodium hydroxide
FarbeWeiß
Schmelzpunkt323°C
SummenformelNaOH
VerpackungMall stakmor case
SynonymSodium hydrate, Soda lye, Caustic soda, Lye
GüteFCC/NF
Molekulargewicht (g/mol)40
Reinheit (%)95.0-100.5%
Menge12 kg
Unit SizeEach