Pierce™ Anti-HA Magnetic Beads, 1 mL - FAQs
Pierce™ Anti-HA Magnetic Beads, 1 mL - FAQs
View additional product information for Pierce™ Anti-HA Magnetic Beads - FAQs (88837, 88836)
1 product FAQs found
Are the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) compatible with sodium dodecyl sulfate? If yes, up to which concentration?
Yes, the Pierce Anti-HA Magnetic Beads (Cat. No. 88836, 88837) are compatible with sodium dodecyl sulfate, but the concentration should not exceed 0.5%.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.