Product Image
Applied Biosystems™
HT WT PLUS Consumables for Biomek™ FXP Target Prep Express
The HT WT PLUS Consumables for Biomek™ FXP Target Prep Express is designed for automation of the target preparation processRead more
Have Questions?
Catalog NumberQuantity
9025081 kit
Catalog number 902508
Price (USD)
2,425.00
Each
Quantity:
1 kit
Price (USD)
2,425.00
Each
The HT WT PLUS Consumables for Biomek™ FXP Target Prep Express is designed for automation of the target preparation process for expression analysis on Applied Biosystems whole-transcriptome (WT) arrays. Designed with reagent chemistries identical to Applied Biosystems GeneChip WT PLUS Reagent Kit, this high-throughput (HT) solution allows the user to generate hybridization-ready targets from input RNA with minimal hands-on time. Used in combination with the Beckman Coulter™ Biomek™ FXP Target Prep Express (TPE) System, HT WT PLUS Kit is capable of processing up to 4x24 or 4x96 reactions in a single run.
Features and benefits
• A complete target prep kit with proven sensitivity and specificity due to an optimized chemistry for Applied Biosystems WT arrays
• Automated protocol that enables the processing of up to 96 samples with minimal hands-on time
• Ability to run a variety of sample types, including clinically relevant samples such as whole blood, to enable robust biomarker identification
• Low sample input requirement (50 ng), allowing you to save your precious samples for further downstream studies
• No requirement for ribosomal RNA depletion or a globin reduction step for blood samples
Related Links
GeneChip™ HT WT PLUS Reagent Kit
GeneChip™ WT PLUS Reagent Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Biomek FXP Target Prep Express
Quantity1 kit
TypeHT WT PLUS Consumables
Unit SizeEach
Frequently asked questions (FAQs)
Is there a recommended RIN (RNA integrity number), based on Bioanalyzer, for RNA quality for IVT and WT assays?
For IVT assays, the RIN should be 6 or higher. For WT assays, there are no recommendations based on assay priming.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.