Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™
Thermo Scientific™

Orion™ Single Junction Refillable Epoxy Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector Types
9157BNMDMini-DIN
9157BNBNC
Catalog number 9157BNMD
Price (MXN)
-
Connector Types:
Mini-DIN
Thermo Scientific™ Refillable Orion™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes have an built-in ATC. They are engineered with an epoxy body for ruggedness and best suited for general purpose samples.
 

Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.

Solutions required

Electrode storage solution

  • 910001 pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH electrode storage solution, 475ml, and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Warranty period

  • 6 months from date of purchase
Specifications
Descriptionmini DIN connector; Glass fiber junction; Refillable
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
For Use With (Application)ideal for field or plant use.
For Use With (Equipment)Orion Star Series Meters
Internal ReferenceAg/AgCl
Length (English)4.7 in.
Length (Metric)120 mm
Temperature (English) Operating32°F to 194°F
Connector TypesMini-DIN
JunctionGlass Fiber
MaterialEpoxy
Temperature (Metric) Operating0°C to 90°C
TypepH/ATC Probe, Mini-DIN connector
pH Range0 to 14
Unit SizeEach