Hydrochloric acid, 36.5-38.0%, NF, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Hydrochloric acid, 36.5-38.0%, NF, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
500 mL
2.5 L
47 lb.
140 lb.
475 lb.
Verpackung:
Alle anzeigen
Clear glass bottle
Clear glass bottle, single shipper
Clear glass bottle, PVC-coated
Katalognummer 9544-03
Preis (EUR)
-
Menge:
2.5 L
Verpackung:
Clear glass bottle
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
This product is GMP Manufactured and meets JP, BP, USP, and EP Requirements.
Specifications
Siedepunkt110°C (1013 hPa)
CAS7647-01-0
FarbeFarblos
Schmelzpunkt-30°C
SummenformelHCl
VerpackungClear glass bottle
SynonymMuriatic Acid, Hydrogen Chloride, aqueous
GüteMulti-Kompendium/NF
Molekulargewicht (g/mol)36.46
Reinheit (%)36.5-38.0%
Menge2.5 L
Unit SizeEach