Thermo Scientific Chemicals

Pentaethylene glycol, 98+%, Thermo Scientific Chemicals

Catalog number: A10133.18
50 g, Each
Thermo Scientific Chemicals

Pentaethylene glycol, 98+%, Thermo Scientific Chemicals

Catalog number: A10133.18
50 g, Each
Quantity
Catalog number: A10133.18
also known as A10133-18
Price (USD)
Price: 483.00
Online price: 417.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4792-15-8
IUPAC Name
3,6,9,12-tetraoxatetradecane-1,14-diol
Molecular Formula
C10H22O6
InChI Key
JLFNLZLINWHATN-UHFFFAOYSA-N
SMILES
OCCOCCOCCOCCOCCO
Appearance (Color)
Clear, colourless to pale yellow
Form
liquid
Assay (Silylated GC)
≥98.0%
Refractive Index
1.4600 - 1.4640 @20?C

Description

Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaethylene glycol is used as an intermediate in the synthesis of cyclic and noncyclic polyethers, complexing agents and immobilized phase-transfer catalysts. It acts as a catalyst for potassium hydroxide induced elimination reactions in benzene. Further, it is used in functional fluids and viscosity adjustors.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text