Thermo Scientific Chemicals

Anthracene, 99%, Thermo Scientific Chemicals

Catalog number: A10203.36
500 g, Each
Thermo Scientific Chemicals

Anthracene, 99%, Thermo Scientific Chemicals

Catalog number: A10203.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10203.36
also known as A10203-36
Price (USD)
Price: 676.00
Your price:
Quantity
-

Chemical Identifiers

CAS
120-12-7
IUPAC Name
anthracene
Molecular Formula
C14H10
InChI Key
MWPLVEDNUUSJAV-UHFFFAOYSA-N
SMILES
C1=CC2=CC3=CC=CC=C3C=C2C=C1
Form
crystalline flakes or powder
Comment
As a bulk commodity material, may contain a small quantity of particulate matter from the manufacturing process
Appearance (Color)
White to cream or pale yellow
Assay (HPLC)
> 98.5%

Description

Anthracene is used in the production of the red dyealizarin and other dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthracene is used in the production of the red dyealizarin and other dyes.

Solubility
Soluble in alcohol, methanol (1.8 g/100 g), hexane (0.37 g/100 g). Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. keep container tightly closed. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text