Coffein, 99 %, Thermo Scientific Chemicals
Coffein, 99 %, Thermo Scientific Chemicals
Coffein, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Coffein, 99 %, Thermo Scientific Chemicals

CAS: 58-08-2 | C8H10N4O2 | 194.19 g/mol
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
100 g
250 g
1000 g
Katalognummer A10431.0B
auch als A10431-0B bezeichnet
Preis (EUR)
131,00
Each
Menge:
1000 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
131,00
Each
Chemikalien-Kennzeichnungen
CAS58-08-2
IUPAC Name1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC8H10N4O2
InChI KeyRYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILESCN1C=NC2=C1C(=O)N(C)C(=O)N2C
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.5 to ≤101.5%
Appearance (Color)White to pale cream
Identification (FTIR)Conforms
This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Löslichkeit
Löslich in Pyrrol, Tetrahydrofuran+Wassergemisch und Ethylacetat. Schwer löslich in Wasser und Ethanol.

Hinweise
Kühl lagern. Nicht kompatibel mit starken Oxidationsmitteln.
RUO – Research Use Only

Allgemeine Referenzen:

  1. Elias V. E. F.; Coso, J. D.; Hamouti, N.; Ortega, JF.; Munoz, G.; Guerra J. M.; Rodriguez R. M. Ingestion of a moderately high caffeine dose before exercise increases postexercise energy expenditure. Int. J. Sport Nutr. Exerc. Metab. 2015, 25 (1), 46-53.
  2. Nakayama, F.; Mizuno, K.; Kato, M. Biosynthesis of caffeine underlying the diversity of motif B' methyltransferase. Nat Prod Commun 10 (5), 799-801.