Thermo Scientific Chemicals

Methyl 3-methyl-2-furoate, 98%, Thermo Scientific Chemicals

Catalog number: A10541.09
10 g, Each
Thermo Scientific Chemicals

Methyl 3-methyl-2-furoate, 98%, Thermo Scientific Chemicals

Catalog number: A10541.09
10 g, Each
Quantity
Catalog number: A10541.09
also known as A10541-09
Price (USD)
Price: 212.00
Online price: 184.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6141-57-7
Appearance (Color)
White to pale yellow
Assay (GC)
≥97.5%
Melting Point (clear melt)
31.0-37.0?C
Form
Crystals or powder or crystalline powder or fused/lumpy solid

Description

Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

Solubility
Insoluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text