Thermo Scientific Chemicals

Octanal, 98%, Thermo Scientific Chemicals

Catalog number: A10901.30
250 g, Each
Thermo Scientific Chemicals

Octanal, 98%, Thermo Scientific Chemicals

Catalog number: A10901.30
250 g, Each
Quantity
Catalog number: A10901.30
also known as A10901-30
Price (USD)
Price: 64.40
Online price: 55.65
Your price:
Quantity
-

Chemical Identifiers

CAS
124-13-0
IUPAC Name
octanal
Molecular Formula
C8H16O
InChI Key
NUJGJRNETVAIRJ-UHFFFAOYSA-N
SMILES
CCCCCCCC=O
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Form
Liquid
Refractive Index
1.4175-1.4215 @ 20?C
Free acid (titration)
≤1%

Description

Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanal is an aromatic aldehyde often found in citrus oils. It is used commercially as a component in perfumes and in flavor production for the food industry.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Air and moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text