Thermo Scientific Chemicals

4-Methoxy-3-buten-2-one, tech. 90%, Thermo Scientific Chemicals

Catalog number: A11235.09
10 g, Each
Thermo Scientific Chemicals

4-Methoxy-3-buten-2-one, tech. 90%, Thermo Scientific Chemicals

Catalog number: A11235.09
10 g, Each
Quantity
Catalog number: A11235.09
also known as A11235-09
Price (USD)
Price: 80.20
Online price: 68.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4652-27-1
Appearance (Color)
Clear, colourless to pale yellow
Refractive Index
1.4610 - 1.4690 @20?C
Form
liquid
Assay (GC)
> 88.0%

Description

4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxy-3-buten-2-one is an important precursor for the synthesis of 1-methoxy-3-trimethylsilyloxy-1,3-butadiene and the Danishefsky diene and its equivalent. It is widely used as a reactant in the synthesis of hydroisoquinoline derivatives. It is employed as an intermediate for the synthesis of manzamine.

Solubility
Miscible with tetrahydrofuran, ether, acetonitrile and benzene.

Notes
Keep away from sources of ignition. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text