Thermo Scientific Chemicals

Phenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: A11246.18
50 g, Each
Thermo Scientific Chemicals

Phenylhydrazine, 97%, Thermo Scientific Chemicals

Catalog number: A11246.18
50 g, Each
Quantity
Catalog number: A11246.18
also known as A11246-18
Price (USD)
Price: 25.90
Your price:
Quantity
-

Chemical Identifiers

CAS
100-63-0
IUPAC Name
hydrogen phenylhydrazine chloride
Molecular Formula
C6H9ClN2
InChI Key
JOVOSQBPPZZESK-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NNC1=CC=CC=C1
Appearance (Color)
Yellow to orange to brown
Assay (Non-aqueous acid-base Titration)
≥96.0% (UK-sourced material)
Comment
Material sourced in UK and US
Form
Liquid or fused solid
Assay from Supplier's CofA
≥96.0% (US-sourced material)

Description

Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl hydrazine is involved in Fischer indole synthesis to prepare indoles, which find application as intermediates in the pharmaceuticals particularly for the tryptamine drug. In analytical chemistry, it is used to differentiate and separate sugars by forming phenyhydrazones. It is used as an N-protecting reagent and for cleavage of phthaloyl group.

Solubility
Soluble in dilute acids.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text