Thermo Scientific Chemicals

Phenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11596.36
500 g, Each
Thermo Scientific Chemicals

Phenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog number: A11596.36
500 g, Each
Quantity
Catalog number: A11596.36
also known as A11596-36
Price (USD)
Price: 205.00
Online price: 174.65
Your price:
Quantity
-

Chemical Identifiers

CAS
103-72-0
IUPAC Name
isothiocyanatobenzene
Molecular Formula
C7H5NS
InChI Key
QKFJKGMPGYROCL-UHFFFAOYSA-N
SMILES
S=C=NC1=CC=CC=C1
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Refractive Index
1.6470-1.6520 @ 20?C

Description

Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl isothiocyanate acts as a derivatizing reagent for primary and secondary amines. It is used in sequencing peptides by Edman degradation and in amino acid analyses by HPLC. It is used for derivatizing N-terminal amino acids of proteins for automated sequential analysis. It is a synthon for dithiadiazafulvalenes.

Solubility
Soluble in alcohol, and ether. Insoluble in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text