Thermo Scientific Chemicals

4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11749.18
50 g, Each
Thermo Scientific Chemicals

4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: A11749.18
50 g, Each
Quantity
Catalog number: A11749.18
also known as A11749-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
5728-52-9
IUPAC Name
2-{[1,1'-biphenyl]-4-yl}acetic acid
Molecular Formula
C14H12O2
InChI Key
QRZAKQDHEVVFRX-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(C=C1)C1=CC=CC=C1
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to cream to pale brown
Melting Point (clear melt)
158.0-165.0?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text