Thermo Scientific Chemicals

Chloroacetone, 95%, stab. with 0.1% epoxidized soybean oil, Thermo Scientific Chemicals

Catalog number: A11922.0B
1000 g, Each
Thermo Scientific Chemicals

Chloroacetone, 95%, stab. with 0.1% epoxidized soybean oil, Thermo Scientific Chemicals

Catalog number: A11922.0B
1000 g, Each
Quantity
Catalog number: A11922.0B
also known as A11922-0B
Price (USD)
Price: 274.00
Online price: 236.65
Your price:
Quantity
-

Chemical Identifiers

CAS
78-95-5
IUPAC Name
1-chloropropan-2-one
Molecular Formula
C3H5ClO
InChI Key
BULLHNJGPPOUOX-UHFFFAOYSA-N
SMILES
CC(=O)CCl
Appearance (Color)
Clear colorless to yellow to brown
Assay (GC)
≥94.0%
Refractive Index
1.4310-1.4370 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

Solubility
Soluble in water (124g/L), ether and chloroform.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Do not store together with strong basic or oxidizing materials.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text