2-Bromoacetamid, 98 %, Thermo Scientific Chemicals
2-Bromoacetamid, 98 %, Thermo Scientific Chemicals
2-Bromoacetamid, 98 %, Thermo Scientific Chemicals
2-Bromoacetamid, 98 %, Thermo Scientific Chemicals
Thermo Scientific™

2-Bromoacetamid, 98 %, Thermo Scientific Chemicals

 
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 g
25 g
Katalognummer A11926.06
auch als A11926-06 bezeichnet
Preis (EUR)
41,40
Exklusiv online
48,70
Ersparnis 7,30 (15%)
Each
Zum Warenkorb hinzufügen
Menge:
5 g
Großbestellung oder individuelle Größe anfordern
Preis (EUR)
41,40
Exklusiv online
48,70
Ersparnis 7,30 (15%)
Each
Zum Warenkorb hinzufügen
Chemikalien-Kennzeichnungen
CAS683-57-8
IUPAC Name2-bromoacetamide
Molecular FormulaC2H4BrNO
InChI KeyJUIKUQOUMZUFQT-UHFFFAOYSA-N
SMILESNC(=O)CBr
Mehr anzeigen
SpecificationsSpecification SheetDatenblatt
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Appearance (Color)White to cream
Assay (GC)≥97.5%
Melting Point (clear melt)85.5-91.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only