Thermo Scientific Chemicals

Perfluoroheptanoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12092.14
25 g, Each
Thermo Scientific Chemicals

Perfluoroheptanoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A12092.14
25 g, Each
Quantity
Catalog number: A12092.14
also known as A12092-14
Price (USD)
Price: 193.00
Online price: 164.65
Your price:
Quantity
-

Chemical Identifiers

CAS
375-85-9
IUPAC Name
tridecafluoroheptanoic acid amine
Molecular Formula
C7H4F13NO2
InChI Key
JVZREVRTMWNFME-UHFFFAOYSA-N
SMILES
N.OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Appearance (Color)
White to pale yellow/orange
Assay from Supplier's CofA
≥98.0 to ≤102.0% (Titration, US sourced material only)
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0% (UK sourced material only)
Form
Crystalline powder or fused solid or clear liquid as melt
Identification (FTIR)
Conforms (UK sourced material only)

Description

Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoroheptanoic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field etc.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text