Thermo Scientific Chemicals

L-Serine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12355.22
100 g, Each
Thermo Scientific Chemicals

L-Serine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: A12355.22
100 g, Each
Quantity
Catalog number: A12355.22
also known as A12355-22
Price (USD)
Price: 317.00
Online price: 269.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5680-80-8
IUPAC Name
methyl 2-amino-3-hydroxypropanoate hydrochloridyl
Molecular Formula
C4H9ClNO3
InChI Key
QDXGLJWDLGMECL-UHFFFAOYNA-N
SMILES
[Cl].COC(=O)C(N)CO
Assay (Titration ex Chloride)
> 97.5%
Water Content (Karl Fischer Titration)
< 1.0%
Optical Rotation
+5 ?0.5? (c=2 in methanol)
Appearance (Color)
White
Form
crystalline powder

Description

L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.

Solubility
Soluble in water (50 mg/ml), and methanol

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text