Thermo Scientific Chemicals

(1-Ethoxycarbonylethyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: A13023.14
25 g, Each
Thermo Scientific Chemicals

(1-Ethoxycarbonylethyl)triphenylphosphonium bromide, 97%, Thermo Scientific Chemicals

Catalog number: A13023.14
25 g, Each
Quantity
Catalog number: A13023.14
also known as A13023-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
30018-16-7
IUPAC Name
(1-ethoxy-1-oxopropan-2-yl)triphenylphosphanium bromide
Molecular Formula
C23H24BrO2P
InChI Key
RSYXORMKBUFAMS-UHFFFAOYNA-M
SMILES
[Br-].CCOC(=O)C(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Crystals or powder or crystalline powder
Assay (Titration ex Bromide)
≥96.0 to ≤104.0%
Water Content (Karl Fischer Titration)
≤1.5%
Appearance (Color)
White to pale cream

Description

1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethoxycarbonylethyl)triphenylphosphonium bromide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is widely used as an organophosphine catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Moisture sensitive. Incompatible to strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text